2-Bromo-6-chlorobenzaldehyde 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD160818-5GEA 28.4 EUR
BLDPBD160818-5G BLDPBD160818-25G BLDPBD160818-100G BLDPBD160818-1G BLDPBD160818-10G BLDPBD160818-250MG
2-Bromo-6-chlorobenzaldehyde 97%
2-Bromo-6-chlorobenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₄BrClO
MW: 219,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00266792
CAS Number: 64622-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR