1-(3-Hydroxy-1-azetidinyl)-ethanone 98%

Synonyms: 1-(3-Hydroxyazetidin-1-yl)ethanone

BLDPBD161914-1GEA 85.1 EUR
BLDPBD161914-1G BLDPBD161914-250MG BLDPBD161914-5G
1-(3-Hydroxy-1-azetidinyl)-ethanone 98%
1-(3-Hydroxy-1-azetidinyl)-ethanone

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₉NO₂
MW: 115,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366667
CAS Number: 118972-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR