3-Bromo-N-tert-butylbenzenesulphonamide 98%

Synonyms: 3-Bromo-N-(tert-butyl)benzenesulphonamide

BLDPBD163252-1GEA 55.7 EUR
BLDPBD163252-1G BLDPBD163252-5G BLDPBD163252-25G BLDPBD163252-250MG
3-Bromo-N-tert-butylbenzenesulphonamide 98%
3-Bromo-N-tert-butylbenzenesulphonamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₄BrNO₂S
MW: 292,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363824
CAS Number: 308283-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR