2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulphonamide 97%
|
|
BLDPBD16342-1GEA
907
EUR
BLDPBD16342-1G
BLDPBD16342-100MG
BLDPBD16342-250MG
2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulphonamide 97%
2,4,6-Trimethyl-N-(2-(trifluoromethyl)phenyl)benzenesulphonamide
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₆H₁₆F₃NO₂S MW: 343,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00784362 CAS Number: 313981-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...