4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD164528-5GEA 273 EUR
BLDPBD164528-5G BLDPBD164528-1G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine hydrochloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₁H₂₀BNO₂·ClH
MW: 245,554 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11506076
CAS Number: 1121057-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR