1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%

Synonyms: tert-Butyl 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate

BLDPBD164807-1GEA 241 EUR
BLDPBD164807-1G BLDPBD164807-250MG BLDPBD164807-5G BLDPBD164807-100MG
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₉H₂₇BN₂O₄
MW: 358,24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195073
CAS Number: 864770-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR