Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%

Synonyms: (2R,4R)-Methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride,cis-4-Hydroxy-D-proline methyl ester hydrochloride,(2R,4R)-4-Hydroxy-2-(methoxycarbonyl)pyrrolidine hydrochloride

BD165788-100G BD165788-1G BD165788-10G BD165788-5G BD165788-250MG BD165788-25G
BLDPBD165788-100GEA 385 EUR
BLDPBD165788-100G BLDPBD165788-1G BLDPBD165788-10G BLDPBD165788-5G BLDPBD165788-250MG BLDPBD165788-25G
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride 97%
Methyl (2R,4R)-4-hydroxypyrrolidine-2-carboxylate hydrochloride
Formula: C₆H₁₁NO₃·ClH
MW: 181,617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082536
CAS Number: 114676-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR