7-Benzyloxy-4-chloro-6-methoxyquinoline 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 7-(Benzyloxy)-4-chloro-6-methoxyquinoline

BLDPBD166715-1GEA 45.2 EUR
BLDPBD166715-1G BLDPBD166715-25G BLDPBD166715-250MG BLDPBD166715-5G
7-Benzyloxy-4-chloro-6-methoxyquinoline 98%
7-Benzyloxy-4-chloro-6-methoxyquinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₇H₁₄ClNO₂
MW: 299,752 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12910463
CAS Number: 286371-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR