4-(2-Hydroxyethyl)phenylboronic acid 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: (4-(2-Hydroxyethyl)phenyl)boronic acid

BLDPBD167826-100MGEA 97.7 EUR
BLDPBD167826-100MG BLDPBD167826-5G BLDPBD167826-1G BLDPBD167826-250MG
4-(2-Hydroxyethyl)phenylboronic acid 97%
4-(2-Hydroxyethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₁₁BO₃
MW: 165,982 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03095135
CAS Number: 137756-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR