Methyl-2-fluoro-3-nitrobenzoate 98%

Synonyms: Methyl 2-fluoro-3-nitrobenzoate

BD168190-10G BD168190-25G BD168190-1G BD168190-250MG BD168190-100G BD168190-5G
BLDPBD168190-10GEA 60.9 EUR
BLDPBD168190-10G BLDPBD168190-25G BLDPBD168190-1G BLDPBD168190-250MG BLDPBD168190-100G BLDPBD168190-5G
Methyl-2-fluoro-3-nitrobenzoate 98%
Methyl 2-fluoro-3-nitrobenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆FNO₄
MW: 199,1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566813
CAS Number: 946126-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR