1-Methylimidazole-5-boronic acid pinacol ester 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

BLDPBD168449-250MGEA 28.4 EUR
BLDPBD168449-250MG BLDPBD168449-5G BLDPBD168449-1G
1-Methylimidazole-5-boronic acid pinacol ester 95%
1-Methylimidazole-5-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₇BN₂O₂
MW: 208,065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846349
CAS Number: 942070-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR