2-Hydroxy-4-methoxy-5-sulphobenzophenone 98%

Synonyms: Benzophenone-4,5-Benzoyl-4-hydroxy-2-methoxybenzenesulphonic acid,2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid

BD17071-10G BD17071-25G BD17071-1000G BD17071-500G BD17071-100G
BLDPBD17071-10GEA 7.4 EUR
BLDPBD17071-10G BLDPBD17071-25G BLDPBD17071-1000G BLDPBD17071-500G BLDPBD17071-100G
2-Hydroxy-4-methoxy-5-sulphobenzophenone 98%
2-Hydroxy-4-methoxy-5-sulfobenzophenone

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₁₂O₆S
MW: 308,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR