3',5'-Dibromoacetophenone 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1-(3,5-Dibromophenyl)ethanone

BLDPBD17237-5GEA 75.6 EUR
BLDPBD17237-5G BLDPBD17237-10G BLDPBD17237-100G BLDPBD17237-25G BLDPBD17237-1G BLDPBD17237-250MG
3',5'-Dibromoacetophenone 97%
3',5'-Dibromoacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆Br₂O
MW: 277,9400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078665
CAS Number: 14401-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR