3-(3-Aminopropoxy)benzonitrile 95+%

Supplier: BLD PHARMATECH GMBH

BLDPBD174298-250MGEA 84 EUR
BLDPBD174298-250MG BLDPBD174298-1G BLDPBD174298-100MG
3-(3-Aminopropoxy)benzonitrile 95+%
3-(3-Aminopropoxy)benzonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₁₂N₂O
MW: 176,2151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16495891
CAS Number: 785760-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR