2,4-Dichloro-D-phenylalanine 97%

BLDPBD17624-25GEA 641 EUR
BLDPBD17624-25G BLDPBD17624-5G BLDPBD17624-250MG BLDPBD17624-10G BLDPBD17624-1G
2,4-Dichloro-D-phenylalanine 97%
2,4-Dichloro-D-phenylalanine

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉Cl₂NO₂
MW: 234,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01860873
CAS Number: 114872-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR