Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate 97%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
Ethyl 4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
BLDPBD181021-10GEA
804
EUR
BLDPBD181021-10G
BLDPBD181021-250MG
BLDPBD181021-1G
BLDPBD181021-100MG
BLDPBD181021-5G
Ethyl-4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate 97%
Ethyl 4-bromo-1,5-dimethyl-1H-pyrazole-3-carboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₁₁BrN₂O₂ MW: 247,0891 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00207970 CAS Number: 5775-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...