Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate 95%

Synonyms: Ethyl 5-amino-1-(p-tolyl)-1H-pyrazole-4-carboxylate

BD18491-5G BD18491-1G BD18491-250MG
BLDPBD18491-5GEA 89.3 EUR
BLDPBD18491-5G BLDPBD18491-1G BLDPBD18491-250MG
Ethyl-5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate 95%
Ethyl 5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₃H₁₅N₃O₂
MW: 245,2800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00973936
CAS Number: 15001-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR