(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
|
|
Synonyms:
PyAOP
, (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate
, ((3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
BLDPBD18599-5GEA
23.1
EUR
BLDPBD18599-5G
BLDPBD18599-10G
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(7-Azabenzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formula:
C₁₇H₂₇F₆N₇OP₂ MW: 521,4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03703417 CAS Number: 156311-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...