Benzyl (S)-(+)-glycidyl ether 97%

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

BD18747-25G BD18747-10G BD18747-5G BD18747-1G
BLDPBD18747-25GEA 32.6 EUR
BLDPBD18747-25G BLDPBD18747-10G BLDPBD18747-5G BLDPBD18747-1G
Benzyl (S)-(+)-glycidyl ether 97%
Benzyl (S)-(+)-glycidyl ether

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054428
CAS Number: 16495-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR