3-(1H-Pyrazol-1-yl)aniline 97%

Synonyms: 3-(1-Pyrazolyl)aniline

BD188085-1G BD188085-250MG BD188085-5G
BLDPBD188085-1GEA 156 EUR
BLDPBD188085-1G BLDPBD188085-250MG BLDPBD188085-5G
3-(1H-Pyrazol-1-yl)aniline 97%
3-(1H-Pyrazol-1-yl)aniline
Formula: C₉H₉N₃
MW: 159,1879 g/mol
MDL Number: MFCD08059554
CAS Number: 184097-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR