4-Amino-3-bromo-2-methoxypyridine 98%

Synonyms: 3-Bromo-2-methoxy-4-pyridinamine3-Bromo-2-methoxypyridin-4-amine

BLDPBD196530-1GEA 69.3 EUR
BLDPBD196530-1G BLDPBD196530-250MG BLDPBD196530-5G BLDPBD196530-25G
4-Amino-3-bromo-2-methoxypyridine 98%
4-Amino-3-bromo-2-methoxypyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₇BrN₂O
MW: 203,0366 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656225
CAS Number: 215364-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR