1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98%

Synonyms: 1,1-Binaphthyl-2,2-diyl hydrogenphosphate

BLDPBD20388-1GEA 7.4 EUR
BLDPBD20388-1G BLDPBD20388-25G BLDPBD20388-5G BLDPBD20388-100G BLDPBD20388-500G
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98%
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₀H₁₃O₄P
MW: 348,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010045
CAS Number: 35193-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR