3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 98%
Supplier: BLD PHARMATECH GMBH
|
|
Synonyms:
3,5-Dimethylpiperidine
BLDPBD20419-10GEA
7.4
EUR
BLDPBD20419-10G
BLDPBD20419-5G
BLDPBD20419-25G
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 98%
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₇H₁₅N MW: 113,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005996 CAS Number: 35794-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...