3-Amino-α,α,α-trifluoro-p-toluic acid 98%
Synonyms:
3-Amino-4-(trifluoromethyl)benzoic acid
BLDPBD20988-1GEA
34.7
EUR
BLDPBD20988-1G
BLDPBD20988-250MG
BLDPBD20988-10G
BLDPBD20988-5G
3-Amino-α,α,α-trifluoro-p-toluic acid 98%
3-Amino-α,α,α-trifluoro-p-toluic acid
Formula:
C₈H₆F₃NO₂ MW: 205,1340 g/mol |
MDL Number:
MFCD07784307 CAS Number: 125483-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...