4-Chloro-2-hydroxybenzonitrile 95%

BD210613-5G BD210613-1G BD210613-25G BD210613-250MG
BLDPBD210613-5GEA 128 EUR
BLDPBD210613-5G BLDPBD210613-1G BLDPBD210613-25G BLDPBD210613-250MG
4-Chloro-2-hydroxybenzonitrile 95%
4-Chloro-2-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153,5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234255
CAS Number: 30818-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR