6-Bromo-3-isoquinolinol 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 6-Bromo-3-hydroxyisoquinoline

BLDPBD210657-250MGEA 17.9 EUR
BLDPBD210657-250MG BLDPBD210657-10G BLDPBD210657-1G BLDPBD210657-25G BLDPBD210657-5G
6-Bromo-3-isoquinolinol 98%
6-Bromo-3-isoquinolinol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₆BrNO
MW: 224,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110423
CAS Number: 1031927-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR