7-Bromoquinoline-3-carboxylic acid 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD211559-100MGEA 32.6 EUR
BLDPBD211559-100MG BLDPBD211559-5G BLDPBD211559-250MG BLDPBD211559-1G
7-Bromoquinoline-3-carboxylic acid 95%
7-Bromoquinoline-3-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₆BrNO₂
MW: 252,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08437554
CAS Number: 892874-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR