3-Bromo-5-cyanobenzaldehyde 98%

Synonyms: 3-Bromo-5-formylbenzonitrile

BLDPBD211788-250MGEA 142 EUR
BLDPBD211788-250MG
3-Bromo-5-cyanobenzaldehyde 98%
3-Bromo-5-cyanobenzaldehyde

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₄BrNO
MW: 210,0275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261040
CAS Number: 644982-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR