5-Bromo-2-chloroquinoline 95%
Supplier: BLD PHARMATECH GMBH
|
|
BLDPBD212313-5GEA
577
EUR
BLDPBD212313-5G
BLDPBD212313-250MG
BLDPBD212313-1G
5-Bromo-2-chloroquinoline 95%
5-Bromo-2-chloroquinoline
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₅BrClN MW: 242,4997 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08437594 CAS Number: 99455-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...