2,6-Dimethylpyridine-3-boronic acid pinacol ester 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2,6-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

BLDPBD213114-250MGEA 92.4 EUR
BLDPBD213114-250MG BLDPBD213114-1G BLDPBD213114-100MG
2,6-Dimethylpyridine-3-boronic acid pinacol ester 95%
2,6-Dimethylpyridine-3-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₂₀BNO₂
MW: 233,114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13195066
CAS Number: 693774-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR