Methyl-3-fluoro-2-nitrobenzoate 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: Methyl 3-fluoro-2-nitrobenzoate

BLDPBD215296-1GEA 52.5 EUR
BLDPBD215296-1G BLDPBD215296-100G BLDPBD215296-25G BLDPBD215296-250MG BLDPBD215296-10G BLDPBD215296-5G
Methyl-3-fluoro-2-nitrobenzoate 98%
Methyl 3-fluoro-2-nitrobenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆FNO₄
MW: 199,1360 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520393
CAS Number: 1214353-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR