4-Amino-2-methoxy-5-nitropyridine 95%

Synonyms: 2-Methoxy-5-nitropyridin-4-amine

BLDPBD215433-100MGEA 76.7 EUR
BLDPBD215433-100MG BLDPBD215433-250MG BLDPBD215433-1G BLDPBD215433-5G
4-Amino-2-methoxy-5-nitropyridine 95%
4-Amino-2-methoxy-5-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₇N₃O₃
MW: 169,138 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977410
CAS Number: 127356-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR