2-Amino-4-cyclopropylpyridine 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4-Cyclopropylpyridin-2-amine

BLDPBD216119-5GEA 934 EUR
BLDPBD216119-5G BLDPBD216119-250MG BLDPBD216119-1G BLDPBD216119-100MG
2-Amino-4-cyclopropylpyridine 95%
2-Amino-4-cyclopropylpyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₁₀N₂
MW: 134,178 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032793
CAS Number: 908269-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR