2-Bromo-3'-(trifluoromethoxy)acetophenone 97%

Synonyms: 3-(Trifluoromethoxy)phenacyl bromide2-Bromo-3'-trifluoromethoxyacetophenone

BLDPBD217425-250MGEA 32.6 EUR
BLDPBD217425-250MG BLDPBD217425-1G BLDPBD217425-5G
2-Bromo-3'-(trifluoromethoxy)acetophenone 97%
2-Bromo-3'-(trifluoromethoxy)acetophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆BrF₃O₂
MW: 283,0400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094425
CAS Number: 237386-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR