2',4'-Dichlorovalerophenone 98+%

Synonyms: 1-(2,4-Dichlorophenyl)pentan-1-one

BD21757-100G BD21757-25G BD21757-5G
BLDPBD21757-100GEA 225 EUR
BLDPBD21757-100G BLDPBD21757-25G BLDPBD21757-5G
2',4'-Dichlorovalerophenone 98+%
2',4'-Dichlorovalerophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂Cl₂O
MW: 231,1184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028061
CAS Number: 61023-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR