1,3-Dibromo-5-isopropylbenzene 96%

BLDPBD217725-25GEA 157 EUR
BLDPBD217725-25G BLDPBD217725-5G BLDPBD217725-1G
1,3-Dibromo-5-isopropylbenzene 96%
1,3-Dibromo-5-isopropylbenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀Br₂
MW: 277,9837 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06657957
CAS Number: 62655-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR