2-Bromo-5-iodobenzaldehyde 97%
|
|
BLDPBD217836-1GEA
65.1
EUR
BLDPBD217836-1G
BLDPBD217836-25G
BLDPBD217836-250MG
BLDPBD217836-5G
BLDPBD217836-10G
2-Bromo-5-iodobenzaldehyde 97%
2-Bromo-5-iodobenzaldehyde
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₄BrIO MW: 310,915 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07779027 CAS Number: 1032231-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...