2-Chloro-6-fluoro-3-methoxybenzoic acid 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD217853-5GEA 343 EUR
BLDPBD217853-5G BLDPBD217853-250MG BLDPBD217853-1G
2-Chloro-6-fluoro-3-methoxybenzoic acid 98%
2-Chloro-6-fluoro-3-methoxybenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆ClFO₃
MW: 204,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115942
CAS Number: 886499-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR