3-Fluoro-2-formylphenylboronic acid 98%

Synonyms: (3-Fluoro-2-formylphenyl)boronic acid3-Fluoro-2-formylbenzeneboronic acid

BLDPBD217988-10GEA 35.7 EUR
BLDPBD217988-10G BLDPBD217988-5G BLDPBD217988-25G BLDPBD217988-1G BLDPBD217988-100G
3-Fluoro-2-formylphenylboronic acid 98%
3-Fluoro-2-formylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₆BFO₃
MW: 167,93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697421
CAS Number: 871126-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR