4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4-(3-Fluorophenylcarbamoyl)benzeneboronic acid(4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid

BLDPBD218006-1GEA 57.8 EUR
BLDPBD218006-1G BLDPBD218006-5G BLDPBD218006-25G
4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₁BFNO₃
MW: 259,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09027204
CAS Number: 874288-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR