4-Bromo-1-naphthaleneboronic acid 97%

Synonyms: (4-Bromonaphthalen-1-yl)boronic acid

BD218038-100MG BD218038-1G BD218038-25G BD218038-250MG BD218038-5G BD218038-10G
BLDPBD218038-100MGEA 6.3 EUR
BLDPBD218038-100MG BLDPBD218038-1G BLDPBD218038-25G BLDPBD218038-250MG BLDPBD218038-5G BLDPBD218038-10G
4-Bromo-1-naphthaleneboronic acid 97%
4-Bromo-1-naphthaleneboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₈BBrO₂
MW: 250,88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701764
CAS Number: 145965-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR