DL-Glutamic acid 97%

BLDPBD21810-100GEA 9.5 EUR
BLDPBD21810-100G BLDPBD21810-25G BLDPBD21810-500G BLDPBD21810-1000G
DL-Glutamic acid 97%
DL-Glutamic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₉NO₄
MW: 147,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063113
CAS Number: 617-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR