2-Amino-2'-fluoroacetophenone hydrochloride 97%
|
|
Synonyms:
2-Fluorophenacylamine hydrochloride
, 2-Amino-1-(2-fluorophenyl)ethanone hydrochloride
BLDPBD218930-250MGEA
10.5
EUR
BLDPBD218930-250MG
BLDPBD218930-1G
2-Amino-2'-fluoroacetophenone hydrochloride 97%
2-Amino-2'-fluoroacetophenone hydrochloride
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₉ClFNO MW: 189,6146 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08532458 CAS Number: 93102-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...