2-Methyl-3-(trifluoromethyl)benzaldehyde 98%

BLDPBD219131-5GEA 139 EUR
BLDPBD219131-5G BLDPBD219131-250MG BLDPBD219131-1G
2-Methyl-3-(trifluoromethyl)benzaldehyde 98%
2-Methyl-3-(trifluoromethyl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₇F₃O
MW: 188,15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04116032
CAS Number: 878001-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR