3-Pyridinesulphonic acid 97%

Synonyms: Pyridine-3-sulphonic acid

BLDPBD21949-10GEA 8.4 EUR
BLDPBD21949-10G BLDPBD21949-5G BLDPBD21949-0.025G BLDPBD21949-25G
3-Pyridinesulphonic acid 97%
3-Pyridinesulphonic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₅NO₃S
MW: 159,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006381
CAS Number: 636-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR