2-Bromo-3-nitroaniline 95%

BLDPBD219682-100MGEA 75.6 EUR
BLDPBD219682-100MG BLDPBD219682-1G BLDPBD219682-250MG BLDPBD219682-5G
2-Bromo-3-nitroaniline 95%
2-Bromo-3-nitroaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅BrN₂O₂
MW: 217,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923157
CAS Number: 35757-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR