Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: Ethyl 2-(1,4-dioxaspiro[4.5]decan-8-ylidene)acetate

BLDPBD220452-5GEA 366 EUR
BLDPBD220452-5G BLDPBD220452-10G BLDPBD220452-250MG BLDPBD220452-1G BLDPBD220452-100MG
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate 95%
Ethyl 1,4-dioxaspiro[4.5]dec-8-ylideneacetate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₈O₄
MW: 226,269 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779434
CAS Number: 51656-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR