N-Benzyloxycarbonyl-L-alaninol 97%

Synonyms: (S)-Benzyl (1-hydroxypropan-2-yl)carbamateN-Cbz-L-Alaninol(S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl esterZ-Ala-ol

BLDPBD22099-10GEA 36.8 EUR
BLDPBD22099-10G BLDPBD22099-5G BLDPBD22099-100G BLDPBD22099-1G BLDPBD22099-25G
N-Benzyloxycarbonyl-L-alaninol 97%
N-Cbz-L-Alaninol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₅NO₃
MW: 209,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672530
CAS Number: 66674-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR