1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%

Synonyms: 1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridine-4-boronic acid,(1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridin-4-yl)boronic acid

BD220991-100MG BD220991-250MG BD220991-5G BD220991-10G BD220991-1G BD220991-25G
BLDPBD220991-100MGEA 39.9 EUR
BLDPBD220991-100MG BLDPBD220991-250MG BLDPBD220991-5G BLDPBD220991-10G BLDPBD220991-1G BLDPBD220991-25G
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₈BNO₄
MW: 227,065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09997795
CAS Number: 844501-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR