N-tert-Butyl-4-boronobenzenesulphonamide 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4-(tert-Butylsulfamoyl)benzeneboronic acid(4-(N-(tert-Butyl)sulfamoyl)phenyl)boronic acid

BLDPBD221147-5GEA 287 EUR
BLDPBD221147-5G BLDPBD221147-250MG BLDPBD221147-1G
N-tert-Butyl-4-boronobenzenesulphonamide 98%
N-tert-Butyl 4-boronobenzenesulphonamide

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₆BNO₄S
MW: 257,11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659841
CAS Number: 208516-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR